Methyl-4-fluoro-5-methoxy-2-nitrobenzoate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 4-fluoro-5-methoxy-2-nitrobenzoate

A814077-1G A814077-250MG A814077-5G
77159-302EA 20 USD
77159-302 77159-304 77159-306
Methyl-4-fluoro-5-methoxy-2-nitrobenzoate 98%
Methyl-4-fluoro-5-methoxy-2-nitrobenzoate

Precaución: for Research Use Only

Formula: C₉H₈FNO₅
MDL Number: MFCD20921594
CAS Number: 159768-50-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR