3-(1,3-Dioxolane)cyclobutanecarboxylic acid 97%

Supplier: Ambeed
Warning

A812988-250MG A812988-1G A812988-5G
77159-300EA 22 USD
77159-300 77893-596 77893-597
3-(1,3-Dioxolane)cyclobutanecarboxylic acid 97%
3-(1,3-Dioxolane)cyclobutanecarboxylic acid

Precaución: for Research Use Only

Formula: C₇H₁₀O₄
MDL Number: MFCD11036133
CAS Number: 1001907-64-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR