4-(4-Chlorophenoxy)benzaldehyde 96%

Supplier: Ambeed
Warning

A811456-1G A811456-25G A811456-5G
77159-290EA 28.84 USD
77159-290 77159-292 77159-294
4-(4-Chlorophenoxy)benzaldehyde 96%
4-(4-Chlorophenoxy)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₃H₉ClO₂
MW: 232.67 g/mol
MDL Number: MFCD01631895
CAS Number: 61343-99-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR