tert-Butyl Hydrogen tetradecanedioate 97%

Supplier: Ambeed
Warning

Synonyms: 14-(tert-Butoxy)-14-oxotetradecanoic acid, Tetradecanedioic acid Mono-tert-butyl ester

A807949-100MG A807949-1G A807949-250MG
77159-252EA 5 USD
77159-252 77159-254 77159-256
tert-Butyl Hydrogen tetradecanedioate 97%
tert-Butyl Hydrogen tetradecanedioate

Precaución: for Research Use Only

Formula: C₁₈H₃₄O₄
MDL Number: MFCD23136040
CAS Number: 234082-00-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR