Tetrakis[N-phthaloyl-(S)-tert-leucinato]dirhodium Bis(ethyl Acetate) Adduct 98%

Supplier: Ambeed
Warning

A807097-100MG A807097-50MG
77159-250EA 150 USD
77159-250 77686-835
Tetrakis[N-phthaloyl-(S)-tert-leucinato]dirhodium Bis(ethyl Acetate) Adduct 98%
Tetrakis[N-phthaloyl-(S)-tert-leucinato]dirhodium Bis(ethyl Acetate) Adduct

Precaución: for Research Use Only

Formula: C₅₆H₅₆N₄O₁₆Rh₂
MDL Number: MFCD06797190
CAS Number: 154090-43-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR