N-Boc-N-methyl-1,3-diaminopropane 97%

Supplier: Ambeed
Warning

Synonyms: 3-(N-Boc-N-methylamino)propylamine, N-Methylpropane-1,3-diamine, N-BOC protected

A806920-1G A806920-250MG A806920-5G
77159-244EA 16.48 USD
77159-244 77159-246 77159-248
N-Boc-N-methyl-1,3-diaminopropane 97%
N-Boc-N-methyl-1,3-diaminopropane

Precaución: for Research Use Only

Formula: C₉H₂₀N₂O₂
MW: 188.271 g/mol
MDL Number: MFCD01631189
CAS Number: 150349-36-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR