BIX 01294 trihydrochloride 99+%

Supplier: Ambeed
Warning

A802290-10MG A802290-1MG A802290-25MG A802290-5MG
77159-230EA 54.59 USD
77159-230 77159-232 77159-234 77159-236
BIX 01294 trihydrochloride 99+%
BIX 01294 trihydrochloride

Precaución: for Research Use Only

Formula: C₂₈H₃₈N₆O₂·3HCl
MDL Number: MFCD16618384
CAS Number: 1392399-03-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR