5-Bromo-2-chlorobenzoyl chloride 97%

Supplier: Ambeed
Danger

Synonyms: 5-Bromo-2-chloro-benzoyl chloride

A801094-10G A801094-1G A801094-25G
77159-224EA 105 USD
77159-224 77159-226 77159-228
5-Bromo-2-chlorobenzoyl chloride 97%
5-Bromo-2-chlorobenzoyl chloride

Precaución: for Research Use Only

Formula: C₇H₃BrCl₂O
MW: 253.92 g/mol
MDL Number: MFCD03208978
CAS Number: 21900-52-7
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR