(S)-1-Methylpyrrolidine-2-carboxamide 97%

Supplier: Ambeed
Warning

A799515-100MG A799515-250MG
77159-220EA 97.85 USD
77159-220 77159-222
(S)-1-Methylpyrrolidine-2-carboxamide 97%
(S)-1-Methylpyrrolidine-2-carboxamide

Precaución: for Research Use Only

Formula: C₆H₁₂N₂O
MW: 128.17 g/mol
MDL Number: MFCD19206880
CAS Number: 94391-71-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR