(1R,2R)-rel-2-(Methoxycarbonyl)cyclopropanecarboxylic acid 98%

Supplier: Ambeed
Warning

A797637-250MG A797637-1G
77159-218EA 76.22 USD
77159-218 77614-424
(1R,2R)-rel-2-(Methoxycarbonyl)cyclopropanecarboxylic acid 98%
(1R,2R)-rel-2-(Methoxycarbonyl)cyclopropanecarboxylic acid

Precaución: for Research Use Only

Formula: C₆H₈O₄
MW: 144.12 g/mol
MDL Number: MFCD29037487
CAS Number: 52920-02-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR