4-(3-Fluorophenyl)-4-oxobutyric acid 98+%

Supplier: Ambeed
Warning

A797442-1G A797442-250MG A797442-5G
77159-210EA 76 USD
77159-210 77159-212 77159-214
4-(3-Fluorophenyl)-4-oxobutyric acid 98+%
4-(3-Fluorophenyl)-4-oxobutyric acid

Precaución: for Research Use Only

Formula: C₁₀H₉FO₃
MDL Number: MFCD01320062
CAS Number: 69797-46-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR