Biotin Alkyne 95%

Supplier: Ambeed
Warning

Synonyms: 5-((3aS,4S,6aR)-2-Oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)-N-(prop-2-yn-1-yl)pentanamide

A797430-250MG A797430-100MG A797430-1G A797430-5G
77159-208EA 41 USD
77159-208 77900-502 77901-268 77901-119
Biotin Alkyne 95%
Biotin Alkyne

Precaución: for Research Use Only

Formula: C₁₃H₁₉N₃O₂S
MDL Number: MFCD28385487
CAS Number: 773888-45-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR