3-(1-(4-Aminobutyl)-1H-1,2,3-triazol-4-yl)aniline trihydrochloride 98+%

Supplier: Ambeed
Warning

Synonyms: 3-[1-(4-aminobutyl)-1h-1,2,3-triazol-4-yl]aniline trihydrochloride

A795208-100G A795208-1G A795208-250MG A795208-25G A795208-5G
77159-190EA 803.4 USD
77159-190 77159-192 77159-194 77159-196 77159-198
3-(1-(4-Aminobutyl)-1H-1,2,3-triazol-4-yl)aniline trihydrochloride 98+%
3-(1-(4-Aminobutyl)-1H-1,2,3-triazol-4-yl)aniline trihydrochloride

Precaución: for Research Use Only

Formula: C₁₂H₂₀Cl₃N₅
MW: 340.67 g/mol
MDL Number: MFCD28502706
CAS Number: 1788044-10-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR