Ethyl 6-methyl-5-nitropyridine-3-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Ethyl 6-methyl-5-nitronicotinate

A794244-1G A794244-250MG A794244-25G A794244-5G
77159-182EA 57.68 USD
77159-182 77159-184 77159-186 77159-188
Ethyl 6-methyl-5-nitropyridine-3-carboxylate 95%
Ethyl 6-methyl-5-nitropyridine-3-carboxylate

Precaución: for Research Use Only

Formula: C₉H₁₀N₂O₄
MW: 210.187 g/mol
MDL Number: MFCD18260802
CAS Number: 1211538-09-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR