4-(1-Methyl-4-piperidyl)phenylboronic Acid Pinacol Ester 98%

Supplier: Ambeed
Warning

A793706-1G A793706-25G A793706-5G
77159-180EA 81 USD
77159-180 77900-938 77901-197
4-(1-Methyl-4-piperidyl)phenylboronic Acid Pinacol Ester 98%
4-(1-Methyl-4-piperidyl)phenylboronic Acid Pinacol Ester

Precaución: for Research Use Only

Formula: C₁₈H₂₈BNO₂
MDL Number: MFCD18383496
CAS Number: 1247000-92-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR