2,4-Difluoro-5-methoxybenzaldehyde 97%
Supplier: Ambeed
|
Warning
|
A792940-100MG
A792940-250MG
A792940-5G
77159-168EA
63.86
USD
77159-168
77159-170
77159-172
2,4-Difluoro-5-methoxybenzaldehyde 97%
2,4-Difluoro-5-methoxybenzaldehyde
Precaución: for Research Use Only
Formula:
C₈H₆F₂O₂ MW: 172.129 g/mol |
MDL Number:
MFCD24539280 CAS Number: 177034-25-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...