4,4'-(4-Nitro-1,3-phenylene)dimorpholine 97%

Supplier: Ambeed
Warning

A792706-1G A792706-250MG A792706-5G
77159-162EA 77.25 USD
77159-162 77159-164 77159-166
4,4'-(4-Nitro-1,3-phenylene)dimorpholine 97%
4,4'-(4-Nitro-1,3-phenylene)dimorpholine

Precaución: for Research Use Only

Formula: C₁₄H₁₉N₃O₄
MW: 293.31 g/mol
MDL Number: MFCD00546019
CAS Number: 133387-34-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR