6,7-Dimethoxyquinolin-4-amine 97%

Supplier: Ambeed
Warning

A792383-100MG A792383-1G A792383-250MG
77159-156EA 93.73 USD
77159-156 77159-158 77159-160
6,7-Dimethoxyquinolin-4-amine 97%
6,7-Dimethoxyquinolin-4-amine

Precaución: for Research Use Only

Formula: C₁₁H₁₂N₂O₂
MW: 204.23 g/mol
MDL Number: MFCD00865929
CAS Number: 13425-92-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR