5-Bromopyrrolo[2,1-f][1,2,4]triazin-4(3H)-one 95%
Supplier: Ambeed
|
Warning
|
A790654-100MG
A790654-250MG
77159-144EA
43
USD
77159-144
77159-146
5-Bromopyrrolo[2,1-f][1,2,4]triazin-4(3H)-one 95%
5-Bromopyrrolo[2,1-f][1,2,4]triazin-4(3H)-one
Precaución: for Research Use Only
Formula:
C₆H₄BrN₃O MW: 214.01 g/mol |
MDL Number:
MFCD23106342 CAS Number: 1403767-05-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...