Boc-1-aminocyclohexane-1-carboxylic acid 98%

Supplier: Ambeed
Warning

A789666-1G A789666-250MG A789666-25G A789666-5G
77159-126EA 14 USD
77159-126 77159-128 77893-553 77893-554
Boc-1-aminocyclohexane-1-carboxylic acid 98%
Boc-1-aminocyclohexane-1-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₂H₂₁NO₄
MDL Number: MFCD00273427
CAS Number: 115951-16-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR