N-(tert-Butoxycarbonylamino)phthalimide 95%

Supplier: Ambeed
Warning

A788497-1G A788497-25G A788497-5G
77159-116EA 15.45 USD
77159-116 77159-118 77159-120
N-(tert-Butoxycarbonylamino)phthalimide 95%
N-(tert-Butoxycarbonylamino)phthalimide

Precaución: for Research Use Only

Formula: C₁₃H₁₄N₂O₄
MW: 262.26 g/mol
MDL Number: MFCD00686624
CAS Number: 34387-89-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR