4-(Furfurylcarbamoyl)phenylboronic acid pinacol ester 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
N-(Furan-2-ylmethyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide, 4-(Furfurylaminocarbonyl)benzeneboronic acid pinacol ester
A788047-100MG
A788047-1G
A788047-250MG
A788047-5G
77159-108EA
11
USD
77159-108
77159-110
77159-112
77159-114
4-(Furfurylcarbamoyl)phenylboronic acid pinacol ester 98%
4-(Furfurylcarbamoyl)phenylboronic acid pinacol ester
Precaución: for Research Use Only
Formula:
C₁₈H₂₂BNO₄ |
MDL Number:
MFCD09266168 CAS Number: 1073353-59-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...