Methyl 3-methyl-1H-pyrazolo[3,4-b]pyridine-5-carboxylate 97%

Supplier: Ambeed
Warning

A787843-100MG A787843-1G A787843-250MG
77159-102EA 59.74 USD
77159-102 77159-104 77159-106
Methyl 3-methyl-1H-pyrazolo[3,4-b]pyridine-5-carboxylate 97%
Methyl 3-methyl-1H-pyrazolo[3,4-b]pyridine-5-carboxylate

Precaución: for Research Use Only

Formula: C₉H₉N₃O₂
MW: 191.18 g/mol
MDL Number: MFCD12031251
CAS Number: 1150618-05-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR