2-Methylquinoline-5-boronic acid pinacol ester 95%

Supplier: Ambeed
Warning

Synonyms: 2-Methylquinoline-5-boronic Acid Pinacol Ester

A786470-100MG A786470-5G
77159-096EA 94.76 USD
77159-096 77159-098
2-Methylquinoline-5-boronic acid pinacol ester 95%
2-Methylquinoline-5-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₆H₂₀BNO₂
MW: 269.146 g/mol
MDL Number: MFCD22418301
CAS Number: 876922-75-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR