Ethyl 3-chloro-1-methyl-1H-pyrazole-5-carboxylate 95%

Supplier: Ambeed
Danger

A783277-100MG A783277-250MG
77159-088EA 32.96 USD
77159-088 77159-090
Ethyl 3-chloro-1-methyl-1H-pyrazole-5-carboxylate 95%
Ethyl 3-chloro-1-methyl-1H-pyrazole-5-carboxylate

Precaución: for Research Use Only

Formula: C₇H₉ClN₂O₂
MW: 188.612 g/mol
MDL Number: MFCD19704132
CAS Number: 173841-07-1
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR