PI3K-IN-1 98+%

Supplier: Ambeed
Warning

A781923-10MG A781923-1MG A781923-2MG A781923-50MG A781923-5MG
77159-072EA 39 USD
77159-072 77159-074 77159-076 77159-078 77159-080
PI3K-IN-1 98+%
PI3K-IN-1

Precaución: for Research Use Only

Formula: C₃₁H₂₉N₅O₆S
MDL Number: MFCD18252649
CAS Number: 1349796-36-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR