Fmoc-O-tert-butyl-D-trans-4-hydroxyproline 98%

Supplier: Ambeed
Warning

Synonyms: Fmoc-trans-D-Hyp(tBu)-OH

A779479-250MG A779479-1G
77159-070EA 170.98 USD
77159-070 77552-420
Fmoc-O-tert-butyl-D-trans-4-hydroxyproline 98%
Fmoc-O-tert-butyl-D-trans-4-hydroxyproline

Precaución: for Research Use Only

Formula: C₂₄H₂₇NO₅
MW: 409.48 g/mol
MDL Number: MFCD11615628
CAS Number: 268729-12-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR