N-(4-Methyl-2-pyridyl)thiourea 95+%

Supplier: Ambeed
Danger

Synonyms: 1-(4-Methylpyridin-2-yl)thiourea

A778501-1G A778501-250MG A778501-5G
77159-062EA 61.8 USD
77159-062 77159-064 77159-066
N-(4-Methyl-2-pyridyl)thiourea 95+%
N-(4-Methyl-2-pyridyl)thiourea

Precaución: for Research Use Only

Formula: C₇H₉N₃S
MW: 167.23 g/mol
MDL Number: MFCD04117348
CAS Number: 21242-21-7
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR