1-Butyl-1-methylpyrrolidinium bromide 97%

Supplier: Ambeed
Warning

Synonyms: N-Butyl-N-methylpyrrolidinium bromide

A774289-100G A774289-500G A774289-5G
77159-050EA 108.15 USD
77159-050 77159-052 77159-054
1-Butyl-1-methylpyrrolidinium bromide 97%
1-Butyl-1-methylpyrrolidinium bromide

Precaución: for Research Use Only

Formula: C₉H₂₀BrN
MW: 222.17 g/mol
MDL Number: MFCD07784439
CAS Number: 93457-69-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR