6-Bromo-1H-pyrrolo[3,2-b]pyridine-2-carboxylic acid 97%

Supplier: Ambeed
Warning

A773230-100MG A773230-1G
77159-038EA 109.18 USD
77159-038 77159-040
6-Bromo-1H-pyrrolo[3,2-b]pyridine-2-carboxylic acid 97%
6-Bromo-1H-pyrrolo[3,2-b]pyridine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₅BrN₂O₂
MDL Number: MFCD18380938
CAS Number: 1260386-47-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR