4-Bromo-3-chloro-2-fluorobenzonitrile 98%

Supplier: Ambeed
Warning

A773021-1G A773021-250MG
77159-034EA 187.46 USD
77159-034 77159-036
4-Bromo-3-chloro-2-fluorobenzonitrile 98%
4-Bromo-3-chloro-2-fluorobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₂BrClFN
MDL Number: MFCD11845913
CAS Number: 1160574-68-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR