(8-Fluoroquinolin-3-yl)boronic acid 95+%

Supplier: Ambeed
Warning

A771255-100MG A771255-1G A771255-250MG A771255-5G
77159-014EA 37 USD
77159-014 77159-016 77159-018 77901-715
(8-Fluoroquinolin-3-yl)boronic acid 95+%
(8-Fluoroquinolin-3-yl)boronic acid

Precaución: for Research Use Only

Formula: C₉H₇BFNO₂
MDL Number: MFCD18382879
CAS Number: 1207750-07-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR