4-Iodo-3-methylbenzonitrile 95%

Supplier: Ambeed
Danger

A766122-1G A766122-250MG A766122-5G
77158-986EA 48.41 USD
77158-986 77158-988 77158-990
4-Iodo-3-methylbenzonitrile 95%
4-Iodo-3-methylbenzonitrile

Precaución: for Research Use Only

Formula: C₈H₆IN
MW: 243.0444 g/mol
MDL Number: MFCD09025697
CAS Number: 42872-85-5
UN: 3439
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR