N-Methyl-3-phenoxybenzylamine 97%

Supplier: Ambeed
Danger

A766049-10G A766049-1G
77158-982EA 259.56 USD
77158-982 77158-984
N-Methyl-3-phenoxybenzylamine 97%
N-Methyl-3-phenoxybenzylamine

Precaución: for Research Use Only

Formula: C₁₄H₁₅NO
MW: 213.28 g/mol
MDL Number: MFCD02089413
CAS Number: 129535-78-0
UN: 2922
ADR: 8,6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR