(R)-2,4-Diaminobutanoic acid dihydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: H-D-Dab-OH·2HCl

A763383-1G A763383-250MG
77158-968EA 33 USD
77158-968 77158-970
(R)-2,4-Diaminobutanoic acid dihydrochloride 98%
(R)-2,4-Diaminobutanoic acid dihydrochloride

Precaución: for Research Use Only

Formula: C₄H₁₂Cl₂N₂O₂
MDL Number: MFCD00236954
CAS Number: 127531-11-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR