tert-Butyl 3-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazol-1-yl)azetidine-1-carboxylate 98%
Supplier: Ambeed
|
Warning
|
A762990-100MG
A762990-1G
A762990-250MG
77158-962EA
64.89
USD
77158-962
77158-964
77158-966
tert-Butyl 3-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazol-1-yl)azetidine-1-carboxylate 98%
tert-Butyl 3-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazol-1-yl)azetidine-1-carboxylate
Precaución: for Research Use Only
Formula:
C₁₇H₂₈BN₃O₄ MW: 349.23 g/mol |
MDL Number:
MFCD18383285 CAS Number: 877399-35-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...