2-Oxaadamantan-1-ylmethanol 95+%

Supplier: Ambeed
Warning

A761508-1G A761508-100MG A761508-250MG
77158-960EA 648 USD
77158-960 77900-559 77901-027
2-Oxaadamantan-1-ylmethanol 95+%
2-Oxaadamantan-1-ylmethanol

Precaución: for Research Use Only

Formula: C₁₀H₁₆O₂
MDL Number: MFCD18909759
CAS Number: 1303974-14-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR