1-Methylindolin-5-amine dihydrochloride 96%

Supplier: Ambeed
Warning

A757370-250MG A757370-1G
77158-938EA 52 USD
77158-938 77900-606
1-Methylindolin-5-amine dihydrochloride 96%
1-Methylindolin-5-amine dihydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₄Cl₂N₂
MDL Number: MFCD16547709
CAS Number: 1240527-25-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR