2-(2-Bromophenyl)-4,6-diphenyl-1,3,5-triazine 99%

Supplier: Ambeed
Warning

A757001-10G A757001-1G A757001-25G A757001-5G
77158-916EA 43 USD
77158-916 77158-918 77158-920 77158-922
2-(2-Bromophenyl)-4,6-diphenyl-1,3,5-triazine 99%
2-(2-Bromophenyl)-4,6-diphenyl-1,3,5-triazine

Precaución: for Research Use Only

Formula: C₂₁H₁₄BrN₃
MDL Number: MFCD29047055
CAS Number: 77989-15-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR