2-Hydroxy-4,5-dimethoxybenzaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 4,5-Dimethoxy-2-hydroxybenzaldehyde

A755272-1G A755272-5G
77158-908EA 147.29 USD
77158-908 77158-910
2-Hydroxy-4,5-dimethoxybenzaldehyde 98%
2-Hydroxy-4,5-dimethoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₁₀O₄
MW: 182.173 g/mol
MDL Number: MFCD00598182
CAS Number: 14382-91-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR