2-(2,3-Dimethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 97%

Supplier: Ambeed
Warning

Synonyms: 2,3-Dimethylphenylboronic acid pinacol ester

A755095-1G A755095-250MG A755095-5G
77158-902EA 43 USD
77158-902 77158-904 77158-906
2-(2,3-Dimethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 97%
2-(2,3-Dimethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₄H₂₁BO₂
MDL Number: MFCD18383605
CAS Number: 1232132-73-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR