4-Methyl-2-pyridyl trifluoromethanesulfonate 98+%

Supplier: Ambeed
Danger

A752875-1G A752875-5G
77158-896EA 12 USD
77158-896 77158-898
4-Methyl-2-pyridyl trifluoromethanesulfonate 98+%
4-Methyl-2-pyridyl trifluoromethanesulfonate

Precaución: for Research Use Only

Formula: C₇H₆F₃NO₃S
MDL Number: MFCD11042681
CAS Number: 179260-78-7
UN: 3082
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR