tert-Butyl ((3S,5R)-5-methylpiperidin-3-yl)carbamate 98%

Supplier: Ambeed
Warning

A751371-100MG A751371-250MG
77158-892EA 100.94 USD
77158-892 77158-894
tert-Butyl ((3S,5R)-5-methylpiperidin-3-yl)carbamate 98%
tert-Butyl ((3S,5R)-5-methylpiperidin-3-yl)carbamate

Precaución: for Research Use Only

Formula: C₁₁H₂₂N₂O₂
MW: 214.3 g/mol
MDL Number: MFCD18072962
CAS Number: 1203651-07-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR