Necrosulfonamide 98+%

Supplier: Ambeed
Warning

Synonyms: (E)-N-(4-(N-(3-Methoxypyrazin-2-yl)sulphamoyl)phenyl)-3-(5-nitrothiophen-2-yl)acrylamide

A750842-10MG A750842-1MG A750842-5MG
77158-880EA 40 USD
77158-880 77158-882 77158-884
Necrosulfonamide 98+%
Necrosulfonamide

Precaución: for Research Use Only

Formula: C₁₈H₁₅N₅O₆S₂
MW: 461.47 g/mol
MDL Number: MFCD02370191
CAS Number: 1360614-48-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR