Butyraldehyde dibutyl acetal 98%

Supplier: Ambeed
Danger

Synonyms: 1,1-Dibutoxybutane, Butyraldehyde Dibutyl Acetal

A749573-1G A749573-250MG A749573-25G A749573-5G
77158-866EA 33 USD
77158-866 77158-868 77158-870 77158-872
Butyraldehyde dibutyl acetal 98%
Butyraldehyde dibutyl acetal

Precaución: for Research Use Only

Formula: C₁₂H₂₆O₂
MW: 202.34 g/mol
MDL Number: MFCD00561028
CAS Number: 5921-80-2
UN: 1993
ADR: 3 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR