BAY 41-2272 98%

Supplier: Ambeed
Warning

Synonyms: 5-Cyclopropyl-2-(1-(2-fluorobenzyl)-1H-pyrazolo[3,4-b]pyridin-3-yl)pyrimidin-4-amine

A749144-10MG A749144-1MG A749144-25MG A749144-50MG A749144-5MG
77158-844EA 142.14 USD
77158-844 77158-846 77158-848 77158-850 77158-852
BAY 41-2272 98%
BAY 41-2272

Precaución: for Research Use Only

Formula: C₂₀H₁₇FN₆
MW: 360.3876 g/mol
MDL Number: MFCD06411391
CAS Number: 256376-24-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR