Alectinib hydrochloride 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
9-Ethyl-6,6-dimethyl-8-(4-morpholin-4-ylpiperidin-1-yl)-11-oxo-5H-benzo[B]carbazole-3-carbonitrile,hydrochloride
A748062-10MG
A748062-25MG
A748062-50MG
A748062-5MG
77158-836EA
6.18
USD
77158-836
77158-838
77158-840
77158-842
Alectinib hydrochloride 98%
Alectinib hydrochloride
Precaución: for Research Use Only
Formula:
C₃₀H₃₅ClN₄O₂ MW: 519.08 g/mol |
MDL Number:
MFCD27987893 CAS Number: 1256589-74-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...