2,3,4-Trifluorobenzonitrile 98%

Supplier: Ambeed
Danger

A747389-10G A747389-25G A747389-5G
77158-820EA 11.33 USD
77158-820 77158-822 77158-824
2,3,4-Trifluorobenzonitrile 98%
2,3,4-Trifluorobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₂F₃N
MW: 157.09 g/mol
MDL Number: MFCD00013288
CAS Number: 143879-80-5
UN: 3276
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR