Methyl octanoate 98%

Supplier: Ambeed
Warning

Synonyms: Caprylic acid methyl ester, Octanoic acid methyl ester, Methyl caprylate

A741710-100G A741710-500G
77158-796EA 8.24 USD
77158-796 77158-798
Methyl octanoate 98%
Methyl octanoate

Precaución: for Research Use Only

Formula: C₉H₁₈O₂
MW: 158.24 g/mol
MDL Number: MFCD00009551
CAS Number: 111-11-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR