4-((2-Formylphenoxy)methyl)benzoic acid 96%

Supplier: Ambeed
Warning

A740581-1G A740581-250MG A740581-5G
77158-786EA 61.8 USD
77158-786 77158-788 77158-790
4-((2-Formylphenoxy)methyl)benzoic acid 96%
4-((2-Formylphenoxy)methyl)benzoic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₂O₄
MW: 256.25 g/mol
MDL Number: MFCD02604798
CAS Number: 338994-68-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR